Product Details

SNP ID
rs61743610
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:90612065 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATAATTGTTTCTAGAGTGAATACT[A/G]TATTGAACAGCTCTTAGAAGACACG
Phenotype
MIM: 607863
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DIRAS2 PubMed Links

Gene Details

Gene
DIRAS2
Gene Name
DIRAS family GTPase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017594.4 2153 UTR 3 NP_060064.2

View Full Product Details