Product Details

SNP ID
rs77050040
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:27963097 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGCTCAGCCTGGAATTGCTCTGCC[A/G]CTGCAGAACCCGGTGGGTCACTGGG
Phenotype
MIM: 600297
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CDX2 PubMed Links

Gene Details

Gene
CDX2
Gene Name
caudal type homeobox 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001265.4 1315 UTR 3 NP_001256.3
XM_011534875.2 1315 UTR 3 XP_011533177.1
XM_011534876.2 1315 Intron XP_011533178.1
XM_011534877.2 1315 Intron XP_011533179.1
XM_011534878.2 1315 UTR 3 XP_011533180.1
XM_011534879.2 1315 UTR 3 XP_011533181.1
Gene
LINC00543
Gene Name
long intergenic non-protein coding RNA 543
There are no transcripts associated with this gene.

View Full Product Details