Product Details

SNP ID
rs1000996110
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:78157457 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCATTTGAATCTTCCTGAGACATCT[C/G]ATAAAGTGGATTTCTCTGTCAAAAT
Phenotype
MIM: 601149
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
IDH3A PubMed Links

Gene Details

Gene
IDH3A
Gene Name
isocitrate dehydrogenase 3 (NAD(+)) alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005530.2 Intron NP_005521.1

View Full Product Details