Product Details

SNP ID
rs34396868
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:18589672 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCCTGCAGGAGCTGGAGCAGACG[A/C]GGCTGGGCACGGTGAGGCCACCACC
Phenotype
MIM: 604237
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
C19orf60 PubMed Links

Gene Details

Gene
C19orf60
Gene Name
chromosome 19 open reading frame 60
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001100418.1 480 Silent Mutation AGG,CGG R148R NP_001093888.1
NM_001100419.1 480 Silent Mutation AGG,CGG R126R NP_001093889.1
XM_005259977.1 480 Silent Mutation AGG,CGG R160R XP_005260034.1
Gene
CRLF1
Gene Name
cytokine receptor like factor 1
There are no transcripts associated with this gene.

View Full Product Details