Product Details

SNP ID
rs75533804
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.12:109481804 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTAAATGATTCAGAAGATGGCTACC[A/G]TTTCTCTCCAAGCCTTTTATAAGTT
Phenotype
MIM: 613421 MIM: 608047
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
KCTD10 PubMed Links

Gene Details

Gene
KCTD10
Gene Name
potassium channel tetramerization domain containing 10
There are no transcripts associated with this gene.

Gene
UBE3B
Gene Name
ubiquitin protein ligase E3B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001270449.1 Intron NP_001257378.1
NM_001270450.1 Intron NP_001257379.1
NM_001270451.1 Intron NP_001257380.1
NM_130466.3 Intron NP_569733.2
NM_183415.2 Intron NP_904324.1
XM_005253987.2 Intron XP_005254044.1
XM_006719681.3 Intron XP_006719744.1
XM_006719682.2 Intron XP_006719745.1
XM_011538959.2 Intron XP_011537261.1
XM_011538961.1 Intron XP_011537263.1
XM_017020195.1 Intron XP_016875684.1
XM_017020196.1 Intron XP_016875685.1

View Full Product Details