Product Details

SNP ID
hCV25753497
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.17:40019976 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAGTGACAGGAGGGAGTTCCATGA[G/T]AGTGGGTGAGGTCACACTCTGGGAG
Phenotype
MIM: 138970 MIM: 607000
Polymorphism
G/T, Transversion Substitution
Allele Nomenclature
Literature Links
CSF3 PubMed Links

Gene Details

Gene
CSF3
Gene Name
colony stimulating factor 3
There are no transcripts associated with this gene.

Gene
MED24
Gene Name
mediator complex subunit 24
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001079518.1 Intron NP_001072986.1
NM_001267797.1 Intron NP_001254726.1
NM_014815.3 Intron NP_055630.2
Gene
MIR6884
Gene Name
microRNA 6884
There are no transcripts associated with this gene.

Gene
SNORD124
Gene Name
small nucleolar RNA, C/D box 124
There are no transcripts associated with this gene.

View Full Product Details