Product Details

SNP ID
rs35974434
Assay Type
Functionally tested
NCBI dbSNP Submissions
11
Location
Chr.1:108650278 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCTGTTGCCTTTCCTCCATTTTTC[C/T]GGAAGATTCCTATCTGGGTACAGTA
Phenotype
MIM: 612178
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HENMT1 PubMed Links

Gene Details

Gene
HENMT1
Gene Name
HEN1 methyltransferase homolog 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001102592.1 1110 Missense Mutation CAG,CGG Q230R NP_001096062.1
NM_144584.2 1110 Missense Mutation CAG,CGG Q230R NP_653185.2
XM_005270411.1 1110 Missense Mutation CAG,CGG Q238R XP_005270468.1
XM_017000179.1 1110 Missense Mutation CAG,CGG Q74R XP_016855668.1

View Full Product Details