Product Details

SNP ID
rs34752670
Assay Type
Functionally tested
NCBI dbSNP Submissions
20
Location
Chr.1:40489205 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATGCTGGAGAACTACAGCAACTTG[C/G]TGTCAGTGGGTAAGACTTGGCTATC
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ZFP69 PubMed Links

Gene Details

Gene
ZFP69
Gene Name
ZFP69 zinc finger protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320178.1 1338 Missense Mutation CTG,GTG L113V NP_001307107.1
NM_001320179.1 1338 Missense Mutation CTG,GTG L113V NP_001307108.1
NM_198494.2 1338 Missense Mutation CTG,GTG L113V NP_940896.2
XM_005270809.4 1338 Missense Mutation CTG,GTG L113V XP_005270866.1
XM_006710605.3 1338 Missense Mutation CTG,GTG L113V XP_006710668.1
XM_006710606.3 1338 Missense Mutation CTG,GTG L113V XP_006710669.1

View Full Product Details