Product Details
- SNP ID
-
rs4378658
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:48916008 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTGAGTCACTTATGGTGGTGTTGCC[A/G]GGCCCCAGGGTACTTGTTCCCTATT
- Phenotype
-
MIM: 611362
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
LOC105371814
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs142959616] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- LOC105371814
- Gene Name
- uncharacterized LOC105371814
There are no transcripts associated with this gene.
- Gene
- UBE2Z
- Gene Name
- ubiquitin conjugating enzyme E2 Z
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_023079.4 |
|
Intron |
|
|
NP_075567.2 |
View Full Product Details