Product Details

SNP ID
rs114754639
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:80079251 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGCCGGGCCCTCGTAGGGCGAGGGG[A/C]GCTCTGGGAACGGGGCCTGTGCGCA
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
TMEM254 PubMed Links

Gene Details

Gene
TMEM254
Gene Name
transmembrane protein 254
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001270367.1 7 Intron NP_001257296.1
NM_001270368.1 7 Intron NP_001257297.1
NM_001270369.1 7 Intron NP_001257298.1
NM_001270370.1 7 Intron NP_001257299.1
NM_001270371.1 7 Intron NP_001257300.1
NM_001270372.1 7 Intron NP_001257301.1
NM_001270373.1 7 Intron NP_001257302.1
NM_001270374.1 7 UTR 5 NP_001257303.1
NM_025125.3 7 Intron NP_079401.2
XM_006717978.2 7 Intron XP_006718041.1
XM_011540192.2 7 Intron XP_011538494.1
XM_011540193.2 7 Intron XP_011538495.1
XM_017016679.1 7 Intron XP_016872168.1
XM_017016680.1 7 Intron XP_016872169.1
Gene
TMEM254-AS1
Gene Name
TMEM254 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details