Product Details

SNP ID
rs78073833
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:57824144 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATACACTCCTGGTCCTCCTGTATAA[C/T]CCTAGGGACCAAAAGGGAGCTGCTG
Phenotype
MIM: 613397 MIM: 608711 MIM: 613057
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AVIL PubMed Links

Gene Details

Gene
AVIL
Gene Name
advillin
There are no transcripts associated with this gene.

Gene
CTDSP2
Gene Name
CTD small phosphatase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005730.3 Intron NP_005721.3
XM_005268556.1 Intron XP_005268613.1
Gene
MIR26A2
Gene Name
microRNA 26a-2
There are no transcripts associated with this gene.

View Full Product Details