Product Details

SNP ID
rs61741737
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.12:27081981 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
CTGATGACTGACCTGAACCATTTCC[A/G]GAGGGGAGCCGCTGGATAGCTGGAA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C12orf71 PubMed Links

Gene Details

Gene
C12orf71
Gene Name
chromosome 12 open reading frame 71
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080406.1 1084 Missense Mutation CCG,CTG P168L NP_001073875.1
XM_011520847.2 1084 Missense Mutation CCG,CTG P168L XP_011519149.1

View Full Product Details