Product Details

SNP ID
rs112343102
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:60292401 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCCTTGGGGTAAGTTGCAAATCTA[A/G]GGGAAGACCAATGGTGTTGCAAACT
Phenotype
MIM: 606547
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MS4A4A PubMed Links

Gene Details

Gene
MS4A4A
Gene Name
membrane spanning 4-domains A4A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001243266.1 Intron NP_001230195.1
NM_024021.3 Intron NP_076926.2
NM_148975.2 Intron NP_683876.1
XM_017017909.1 Intron XP_016873398.1

View Full Product Details