Product Details

SNP ID
rs34750131
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:89645070 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GATGACAAAGATATTTGCATTCCGG[A/G]CAGCCTGAACTGCTGCCAGGACTCT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC101929057 PubMed Links

Gene Details

Gene
LOC101929057
Gene Name
uncharacterized LOC101929057
There are no transcripts associated with this gene.

Gene
LYRM2
Gene Name
LYR motif containing 2
There are no transcripts associated with this gene.

Gene
MDN1
Gene Name
midasin AAA ATPase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014611.2 16539 Missense Mutation GCC,GTC A5516V NP_055426.1
XM_005248700.3 16539 Missense Mutation GCC,GTC A3511V XP_005248757.1
XM_006715405.3 16539 Missense Mutation GCC,GTC A3664V XP_006715468.1
XM_011535635.2 16539 Missense Mutation GCC,GTC A5419V XP_011533937.1
XM_011535636.2 16539 Intron XP_011533938.1

View Full Product Details