Product Details
- SNP ID
-
rs3767609
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
27
- Location
-
Chr.1:111414895 on Build GRCh38
- Set Membership
-
HapMap
JSNP
- Context Sequence [VIC/FAM]
- CCTCCAGGGCTCACAGACTGATGAC[T/C]CACAGGGGTCACAGACTGATGACCC
- Phenotype
-
MIM: 603578
- Polymorphism
- T/C, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
OVGP1
PubMed Links
Gene Details
- Gene
- OVGP1
- Gene Name
- oviductal glycoprotein 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_002557.3 |
1662 |
Missense Mutation |
AGT,GGT |
S536G |
NP_002548.3 |
View Full Product Details