Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_014641.2 | Intron | NP_055456.2 | |||
XM_005249492.2 | Intron | XP_005249549.1 | |||
XM_005249493.3 | Intron | XP_005249550.1 | |||
XM_005249494.4 | Intron | XP_005249551.1 | |||
XM_005249497.3 | Intron | XP_005249554.1 | |||
XM_005249498.3 | Intron | XP_005249555.1 | |||
XM_011515001.2 | Intron | XP_011513303.1 | |||
XM_011515003.2 | Intron | XP_011513305.1 | |||
XM_011515004.2 | Intron | XP_011513306.1 | |||
XM_017011519.1 | Intron | XP_016867008.1 | |||
XM_017011520.1 | Intron | XP_016867009.1 | |||
XM_017011521.1 | Intron | XP_016867010.1 | |||
XM_017011522.1 | Intron | XP_016867011.1 |