Product Details

SNP ID
rs35231253
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:50218105 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGTTCATGACGGGCGCCGCCTTCTC[C/T]GTGAGCAGGCCCCTGGGGGGAAGCA
Phenotype
MIM: 607917 MIM: 610053
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SELO PubMed Links

Gene Details

Gene
SELO
Gene Name
selenoprotein O
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_031454.1 5693 Intron NP_113642.1
Gene
TUBGCP6
Gene Name
tubulin gamma complex associated protein 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020461.3 5693 Silent Mutation ACA,ACG T1727T NP_065194.2

View Full Product Details