Product Details

SNP ID
rs78680590
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:57474514 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAGGAGAGATCAGGAGCTAAGACA[C/T]ACACTTCCAGCCTGACGGCCTCTCT
Phenotype
MIM: 610576 MIM: 165220
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ARHGAP9 PubMed Links

Gene Details

Gene
ARHGAP9
Gene Name
Rho GTPase activating protein 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080156.2 Intron NP_001073625.1
NM_001080157.1 Intron NP_001073626.1
NM_001319850.1 Intron NP_001306779.1
NM_001319851.1 Intron NP_001306780.1
NM_001319852.1 Intron NP_001306781.1
NM_032496.3 Intron NP_115885.2
XM_005269083.2 Intron XP_005269140.1
XM_005269084.2 Intron XP_005269141.1
XM_005269085.2 Intron XP_005269142.1
XM_011538656.2 Intron XP_011536958.1
XM_011538657.2 Intron XP_011536959.1
XM_011538658.2 Intron XP_011536960.1
XM_011538659.2 Intron XP_011536961.1
XM_017019800.1 Intron XP_016875289.1
XM_017019801.1 Intron XP_016875290.1
XM_017019802.1 Intron XP_016875291.1
XM_017019803.1 Intron XP_016875292.1
XM_017019804.1 Intron XP_016875293.1
Gene
GLI1
Gene Name
GLI family zinc finger 1
There are no transcripts associated with this gene.

View Full Product Details