Product Details
- SNP ID
-
rs17123362
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:48862181 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGTTTTCCTTCCCCAATAGGCCTAA[A/G]CCCTAAAGCCCTGGGAATACCATCC
- Phenotype
-
MIM: 609038
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
RND1
PubMed Links
Gene Details
- Gene
- RND1
- Gene Name
- Rho family GTPase 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_014470.3 |
|
Intron |
|
|
NP_055285.1 |
View Full Product Details