Product Details

SNP ID
rs34815989
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:31712691 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAGTGCCAATACTGATGCCACAAG[C/T]TTCATCGTCCCGACACTCGGTGGGA
Phenotype
MIM: 142620 MIM: 606520 MIM: 610435 MIM: 606038 MIM: 610437 MIM: 611404
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ABHD16A PubMed Links

Gene Details

Gene
ABHD16A
Gene Name
abhydrolase domain containing 16A
There are no transcripts associated with this gene.

Gene
C6orf25
Gene Name
chromosome 6 open reading frame 25
There are no transcripts associated with this gene.

Gene
LY6G6C
Gene Name
lymphocyte antigen 6 complex, locus G6C
There are no transcripts associated with this gene.

Gene
LY6G6D
Gene Name
lymphocyte antigen 6 complex, locus G6D
There are no transcripts associated with this gene.

Gene
LY6G6E
Gene Name
lymphocyte antigen 6 complex, locus G6E (pseudogene)
There are no transcripts associated with this gene.

Gene
LY6G6F
Gene Name
lymphocyte antigen 6 complex, locus G6F
There are no transcripts associated with this gene.

View Full Product Details