Product Details

SNP ID
rs45572335
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:58153724 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACTTGCAAGAATAAAACTAAAGAG[C/T]ATGTTGTAGGCAAAGATCACTAACT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C20orf85 PubMed Links

Gene Details

Gene
C20orf85
Gene Name
chromosome 20 open reading frame 85
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178456.2 Intron NP_848551.1

View Full Product Details