Product Details

SNP ID
rs61733760
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:124522721 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGAACAGGTCATTCCTTGCCCACA[C/T]TGGTCTTGCAGGAATGCAGCACCAC
Phenotype
MIM: 602778
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NR6A1 PubMed Links

Gene Details

Gene
NR6A1
Gene Name
nuclear receptor subfamily 6 group A member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278546.1 1603 Missense Mutation NP_001265475.1
NM_001489.4 1603 Missense Mutation NP_001480.3
NM_033334.3 1603 Missense Mutation NP_201591.2
XM_005251917.4 1603 Missense Mutation XP_005251974.1
XM_005251918.4 1603 Missense Mutation XP_005251975.1

View Full Product Details