Product Details

SNP ID
rs112440368
Assay Type
Functionally tested
NCBI dbSNP Submissions
6
Location
Chr.1:23787179 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGGAGATATTAAATGATGACTCCT[A/C]GAAATGAACCTGAATAAGGACTACC
Phenotype
MIM: 606953 MIM: 616143
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
GALE PubMed Links

Gene Details

Gene
GALE
Gene Name
UDP-galactose-4-epimerase
There are no transcripts associated with this gene.

Gene
LYPLA2
Gene Name
lysophospholipase II
There are no transcripts associated with this gene.

Gene
PITHD1
Gene Name
PITH domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020362.4 Intron NP_065095.2
XM_011541804.1 Intron XP_011540106.1
Gene
TCEB3-AS1
Gene Name
TCEB3 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details