Product Details

SNP ID
rs140950939
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:23127029 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTATGAATGTCAGTTTTTGTTTACA[A/G]TTTCTCACCACCCACACCAAAGTCC
Phenotype
MIM: 600749 MIM: 604235
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CEBPE PubMed Links

Gene Details

Gene
CEBPE
Gene Name
CCAAT/enhancer binding protein epsilon
There are no transcripts associated with this gene.

Gene
SLC7A8
Gene Name
solute carrier family 7 member 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001267036.1 1286 UTR 3 NP_001253965.1
NM_001267037.1 1286 UTR 3 NP_001253966.1
NM_012244.3 1286 UTR 3 NP_036376.2
NM_182728.2 1286 UTR 3 NP_877392.1

View Full Product Details