Product Details

SNP ID
rs3742305
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:30462505 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACAATCATACATCTGGCGTACTTGT[C/G]ATCATTTTACCAAGCAAACCCACAC
Phenotype
MIM: 163905
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
HMGB1 PubMed Links

Gene Details

Gene
HMGB1
Gene Name
high mobility group box 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001313892.1 Intron NP_001300821.1
NM_001313893.1 Intron NP_001300822.1
NM_002128.5 Intron NP_002119.1

View Full Product Details