Product Details

SNP ID
hCV25759220
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:118616297 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAGGGGAGGCTGGCTGTGGTGGTT[C/G]CCATGGTAACTGCCTGAGTGACCAT
Phenotype
MIM: 612834
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
PHLDB1 PubMed Links

Gene Details

Gene
PHLDB1
Gene Name
pleckstrin homology like domain family B member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001144758.2 Intron NP_001138230.1
NM_001144759.2 Intron NP_001138231.1
NM_015157.3 Intron NP_055972.1

View Full Product Details