Product Details

SNP ID
rs1052444671
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.11:77332592 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAAAGGCCCAGGGATAAAAGGGCA[A/G]TAGGAACAGAGTGAGTGTAGGAAGT
Phenotype
MIM: 602590
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
PAK1 PubMed Links

Gene Details

Gene
PAK1
Gene Name
p21 (RAC1) activated kinase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001128620.1 Intron NP_001122092.1
NM_002576.4 Intron NP_002567.3

View Full Product Details