Product Details

SNP ID
rs191774854
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:155511900 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGTTACTCACAAATAACAGTAGAT[A/T]TTTTTGGTTCCAGAATTTAATCCAT
Phenotype
MIM: 300777
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
TMLHE PubMed Links

Gene Details

Gene
TMLHE
Gene Name
trimethyllysine hydroxylase, epsilon
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001184797.1 Intron NP_001171726.1
NM_018196.3 Intron NP_060666.1
XM_011531182.2 Intron XP_011529484.1
XM_017029620.1 Intron XP_016885109.1

View Full Product Details