Product Details
- SNP ID
-
rs61736755
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:73127742 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTTTGAGAGTGTACACTCGGTACTT[A/C]ACCACCAGAGGCAGGCTCTCAATAA
- Phenotype
-
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
NAP1L6
PubMed Links
Gene Details
- Gene
- NAP1L6
- Gene Name
- nucleosome assembly protein 1 like 6
There are no transcripts associated with this gene.
View Full Product Details