Product Details
- SNP ID
-
rs61473705
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.22:50193599 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGGCCAACGTGGAACCTGTTGTGCC[A/G]TCAGAGGCTTCAGAGCCGGTGCCCA
- Phenotype
-
MIM: 607917
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
SELO
PubMed Links
Gene Details
- Gene
- SELO
- Gene Name
- selenoprotein O
There are no transcripts associated with this gene.
- Gene
- TRABD
- Gene Name
- TraB domain containing
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001320484.1 |
190 |
Silent Mutation |
CCA,CCG |
P19P |
NP_001307413.1 |
NM_001320485.1 |
190 |
Silent Mutation |
CCA,CCG |
P19P |
NP_001307414.1 |
NM_001320487.1 |
190 |
Silent Mutation |
CCA,CCG |
P19P |
NP_001307416.1 |
NM_001320488.1 |
190 |
Silent Mutation |
CCA,CCG |
P19P |
NP_001307417.1 |
NM_025204.3 |
190 |
Silent Mutation |
CCA,CCG |
P19P |
NP_079480.2 |
XM_006724422.3 |
190 |
Silent Mutation |
CCA,CCG |
P19P |
XP_006724485.1 |
XM_006724423.3 |
190 |
Silent Mutation |
CCA,CCG |
P19P |
XP_006724486.1 |
XM_011530712.2 |
190 |
Silent Mutation |
CCA,CCG |
P19P |
XP_011529014.2 |
XM_011530713.2 |
190 |
Silent Mutation |
CCA,CCG |
P19P |
XP_011529015.2 |
XM_011530715.2 |
190 |
Silent Mutation |
CCA,CCG |
P19P |
XP_011529017.2 |
XM_011530716.2 |
190 |
Silent Mutation |
CCA,CCG |
P19P |
XP_011529018.1 |
XM_011530717.2 |
190 |
Silent Mutation |
CCA,CCG |
P19P |
XP_011529019.1 |
XM_011530718.2 |
190 |
Silent Mutation |
CCA,CCG |
P19P |
XP_011529020.1 |
XM_017028943.1 |
190 |
Silent Mutation |
CCA,CCG |
P19P |
XP_016884432.1 |
XM_017028944.1 |
190 |
Silent Mutation |
CCA,CCG |
P19P |
XP_016884433.1 |
View Full Product Details