Product Details

SNP ID
rs115850805
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:60173150 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTCCCTGAAAGTCAAGAAATAAAA[C/G]ATTGATGTTGCTTAGGTCAGCTGAA
Phenotype
MIM: 606548
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
MS4A6A PubMed Links

Gene Details

Gene
MS4A6A
Gene Name
membrane spanning 4-domains A6A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001247999.1 Intron NP_001234928.1
NM_022349.3 Intron NP_071744.2
NM_152851.2 Intron NP_690590.1
NM_152852.2 Intron NP_690591.1
XM_005274177.2 Intron XP_005274234.1
XM_006718658.2 Intron XP_006718721.1
XM_006718660.2 Intron XP_006718723.1
XM_006718661.2 Intron XP_006718724.1
XM_011545209.1 Intron XP_011543511.1
XM_017018123.1 Intron XP_016873612.1
XM_017018124.1 Intron XP_016873613.1
XM_017018125.1 Intron XP_016873614.1

View Full Product Details