Product Details

SNP ID
rs113504041
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.20:63493378 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGGGTGCTTCCAGGAGTACTGCT[A/G]TTCAGGCTAAACTTGCCTCGTGCCC
Phenotype
MIM: 602959
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
EEF1A2 PubMed Links

Gene Details

Gene
EEF1A2
Gene Name
eukaryotic translation elongation factor 1 alpha 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001958.3 Intron NP_001949.1

View Full Product Details