Product Details

SNP ID
rs61729386
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:20369944 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTCATGATCGCCTTGACTAGCGC[G/T]GTCAGGACCGTGTCCCTCATTTCCT
Phenotype
MIM: 123805
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
PDE3A PubMed Links

Gene Details

Gene
PDE3A
Gene Name
phosphodiesterase 3A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000921.4 700 Silent Mutation GCG,GCT A220A NP_000912.3
NM_001244683.1 700 Intron NP_001231612.1
XM_017019420.1 700 Silent Mutation GCG,GCT A220A XP_016874909.1
XM_017019421.1 700 Intron XP_016874910.1

View Full Product Details