Product Details

SNP ID
rs1036614054
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:133083364 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTAAGATGGGGACGTAACTGCAC[C/G]TTATTTTTAGACATTTTAAAAACTT
Phenotype
MIM: 604082
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ZNF140 PubMed Links

Gene Details

Gene
ZNF140
Gene Name
zinc finger protein 140
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300776.1 Intron NP_001287705.1
NM_001300777.1 Intron NP_001287706.1
NM_001300778.1 Intron NP_001287707.1
NM_003440.3 Intron NP_003431.2

View Full Product Details