Product Details

SNP ID
rs973787801
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:54969963 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATTTGAGACAGGAGTGCTAATCAC[C/T]ATCCCTCTCCACTCCTCCCTTGATT
Phenotype
MIM: 609364 MIM: 609661
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NLRP2 PubMed Links

Gene Details

Gene
NLRP2
Gene Name
NLR family pyrin domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001174083.1 Intron NP_001167554.1
Gene
NLRP7
Gene Name
NLR family pyrin domain containing 7
There are no transcripts associated with this gene.

View Full Product Details