Product Details

SNP ID
rs184930041
Assay Type
Functionally tested
NCBI dbSNP Submissions
10
Location
Chr.1:236552188 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTAAGAGCTGTACTGACTTGAGACA[A/G]GCTCTAACTTTTTAAACATTAGTTC
Phenotype
MIM: 606099
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HEATR1 PubMed Links

Gene Details

Gene
HEATR1
Gene Name
HEAT repeat containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018072.5 5362 Intron NP_060542.4
Gene
LGALS8
Gene Name
galectin 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006499.4 5362 UTR 3 NP_006490.3
NM_201543.2 5362 UTR 3 NP_963837.1
NM_201544.2 5362 UTR 3 NP_963838.1
NM_201545.2 5362 UTR 3 NP_963839.1
XM_011544188.2 5362 Intron XP_011542490.1
XM_017001274.1 5362 Intron XP_016856763.1
XM_017001275.1 5362 Intron XP_016856764.1

View Full Product Details