Product Details

SNP ID
rs116412969
Assay Type
Functionally tested
NCBI dbSNP Submissions
13
Location
Chr.1:150997720 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAGCTGCTGGGCCAGCTCCAAGTC[A/G]GTAAGCCCCAGCGGGCCTCGTGGCT
Phenotype
MIM: 603319
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ANXA9 PubMed Links

Gene Details

Gene
ANXA9
Gene Name
annexin A9
There are no transcripts associated with this gene.

Gene
FAM63A
Gene Name
family with sequence similarity 63 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001040217.2 1913 Silent Mutation NP_001035307.1
NM_001163258.1 1913 Silent Mutation NP_001156730.1
NM_001163259.1 1913 Silent Mutation NP_001156731.1
NM_001163260.1 1913 Silent Mutation NP_001156732.1
NM_001319998.1 1913 Silent Mutation NP_001306927.1
NM_018379.4 1913 Silent Mutation NP_060849.2
XM_005245321.3 1913 Silent Mutation XP_005245378.1
XM_005245325.3 1913 Silent Mutation XP_005245382.1
XM_005245327.3 1913 Silent Mutation XP_005245384.1
XM_005245328.3 1913 Silent Mutation XP_005245385.1
XM_011509757.2 1913 Silent Mutation XP_011508059.1
XM_011509759.2 1913 Silent Mutation XP_011508061.1
XM_017001770.1 1913 Silent Mutation XP_016857259.1
XM_017001771.1 1913 Silent Mutation XP_016857260.1
XM_017001772.1 1913 Silent Mutation XP_016857261.1
XM_017001773.1 1913 Silent Mutation XP_016857262.1
XM_017001774.1 1913 Silent Mutation XP_016857263.1
XM_017001775.1 1913 Silent Mutation XP_016857264.1
XM_017001776.1 1913 Silent Mutation XP_016857265.1
XM_017001777.1 1913 Silent Mutation XP_016857266.1

View Full Product Details