Product Details

SNP ID
rs11574915
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:34710087 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCCATGTCTGTGGTAGAGGGTGAGT[A/C]AGAGGCCAGAGCTGAGGGTGAGGTG
Phenotype
MIM: 602737
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CCL21 PubMed Links

Gene Details

Gene
CCL21
Gene Name
C-C motif chemokine ligand 21
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002989.3 81 UTR 5 NP_002980.1
XM_011518004.2 81 UTR 5 XP_011516306.1

View Full Product Details