Product Details

SNP ID
rs2076114
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:38113432 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCCCCACAGGATGAGGGGAAGCCAT[C/T]GACCTGGGCTACAGACCCTGAGGGA
Phenotype
MIM: 603604
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BAIAP2L2 PubMed Links

Gene Details

Gene
BAIAP2L2
Gene Name
BAI1 associated protein 2 like 2
There are no transcripts associated with this gene.

Gene
PLA2G6
Gene Name
phospholipase A2 group VI
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004426.1 Intron NP_001004426.1
NM_001199562.1 Intron NP_001186491.1
NM_003560.2 Intron NP_003551.2
XM_005261764.2 Intron XP_005261821.1
XM_005261765.1 Intron XP_005261822.1
XM_005261766.1 Intron XP_005261823.1
XM_006724332.3 Intron XP_006724395.1
XM_011530422.1 Intron XP_011528724.1
XM_011530423.1 Intron XP_011528725.1
XM_011530424.1 Intron XP_011528726.1
XM_011530425.1 Intron XP_011528727.1
XM_011530426.2 Intron XP_011528728.1
XM_017028981.1 Intron XP_016884470.1
XM_017028982.1 Intron XP_016884471.1
XM_017028983.1 Intron XP_016884472.1
XM_017028984.1 Intron XP_016884473.1
XM_017028985.1 Intron XP_016884474.1
XM_017028986.1 Intron XP_016884475.1
XM_017028987.1 Intron XP_016884476.1
XM_017028988.1 Intron XP_016884477.1

View Full Product Details