Product Details

SNP ID
rs4239710
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:13717359 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCTTTAAGAGGCTATGCCTGGTGTC[C/T]ATGGTTACCTCAAAGTCATCCTCTA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ESF1 PubMed Links

Gene Details

Gene
ESF1
Gene Name
ESF1 nucleolar pre-rRNA processing protein homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001276380.1 Intron NP_001263309.1
NM_016649.3 Intron NP_057733.2
XM_017027874.1 Intron XP_016883363.1

View Full Product Details