Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_023944.3 | 161 | Intron | NP_076433.3 | ||
XM_006722850.3 | 161 | UTR 5 | XP_006722913.1 | ||
XM_011528202.2 | 161 | Intron | XP_011526504.1 | ||
XM_011528203.2 | 161 | Intron | XP_011526505.1 | ||
XM_011528204.2 | 161 | Intron | XP_011526506.1 | ||
XM_011528205.2 | 161 | Intron | XP_011526507.1 | ||
XM_011528207.2 | 161 | Intron | XP_011526509.1 | ||
XM_011528208.2 | 161 | Intron | XP_011526510.1 | ||
XM_017027172.1 | 161 | Missense Mutation | ACG,ATG | T16M | XP_016882661.1 |