Product Details

SNP ID
rs61733926
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:46275299 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGAACAGATTCTCCAGGACCACCC[A/G]ACAGACATGTTGGCCCTGAAATTTT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TTC38 PubMed Links

Gene Details

Gene
TTC38
Gene Name
tetratricopeptide repeat domain 38
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017931.2 493 Silent Mutation CCA,CCG P139P NP_060401.2
XM_011530259.1 493 Silent Mutation CCA,CCG P74P XP_011528561.1
XM_011530260.2 493 Silent Mutation CCA,CCG P139P XP_011528562.1

View Full Product Details