Product Details
- SNP ID
-
rs34852828
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:113408878 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAGCCTTTCTTCGTCTTCATCATTG[G/T]CCTCATCCCTGCTCTTGTCTTGCTC
- Phenotype
-
MIM: 125270
MIM: 607267
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
ALAD
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs3827674] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ALAD
- Gene Name
- aminolevulinate dehydratase
There are no transcripts associated with this gene.
- Gene
- C9orf43
- Gene Name
- chromosome 9 open reading frame 43
There are no transcripts associated with this gene.
- Gene
- POLE3
- Gene Name
- DNA polymerase epsilon 3, accessory subunit
View Full Product Details