Product Details

SNP ID
rs12726906
Assay Type
Validated
NCBI dbSNP Submissions
34
Location
Chr.1:3857748 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CGCTGGGCTAGGCGGGGACGGCCCG[T/C]CGGGGAGGCGTGTGGGGAGAATAGG
Phenotype
MIM: 616690 MIM: 601883
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
CEP104 PubMed Links

Gene Details

Gene
CEP104
Gene Name
centrosomal protein 104
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014704.3 Intron NP_055519.1
XM_005244815.4 Intron XP_005244872.1
XM_011542473.2 Intron XP_011540775.1
XM_011542474.2 Intron XP_011540776.1
XM_011542475.2 Intron XP_011540777.1
XM_011542476.2 Intron XP_011540778.1
XM_011542477.2 Intron XP_011540779.1
XM_011542478.1 Intron XP_011540780.1
XM_017002917.1 Intron XP_016858406.1
XM_017002918.1 Intron XP_016858407.1
XM_017002919.1 Intron XP_016858408.1
Gene
DFFB
Gene Name
DNA fragmentation factor subunit beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282669.1 Intron NP_001269598.1
NM_001320132.1 Intron NP_001307061.1
NM_001320136.1 Intron NP_001307065.1
NM_004402.3 Intron NP_004393.1
XM_011540865.2 Intron XP_011539167.1
XM_017000498.1 Intron XP_016855987.1
XM_017000499.1 Intron XP_016855988.1
XM_017000500.1 Intron XP_016855989.1

View Full Product Details