Product Details

SNP ID
rs35806103
Assay Type
Functionally tested
NCBI dbSNP Submissions
25
Location
Chr.1:3022005 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGCGTGAAACCCAGCGACCAGCCC[C/T]TGCTTGCAACGGAGCCCTCCCTGAA
Phenotype
MIM: 608535
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ACTRT2 PubMed Links

Gene Details

Gene
ACTRT2
Gene Name
actin related protein T2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_080431.4 524 Silent Mutation CTG,TTG L107L NP_536356.3

View Full Product Details