Product Details
- SNP ID
-
hCV25764307
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:88803166 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCATACTAGAAGAGCATAGTGATGA[A/C]GTAGCAAATTGTACTGGAAAGAATG
- Phenotype
-
MIM: 613923
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
LIPM
PubMed Links
Gene Details
- Gene
- LIPM
- Gene Name
- lipase family member M
View Full Product Details