Product Details

SNP ID
rs55646457
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:58429536 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGATCCAGAACCTAAGTGACCTGAG[A/T]GGTAAAAAGCTAAGTATGTGAACTG
Phenotype
MIM: 179060 MIM: 611450
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
PDHB PubMed Links

Gene Details

Gene
PDHB
Gene Name
pyruvate dehydrogenase (lipoamide) beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000925.3 Intron NP_000916.2
NM_001173468.1 Intron NP_001166939.1
NM_001315536.1 Intron NP_001302465.1
Gene
PXK
Gene Name
PX domain containing serine/threonine kinase like
There are no transcripts associated with this gene.

View Full Product Details