Product Details
- SNP ID
-
hCV25764991
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:44538648 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATAAAATCTTATGTCTTTGCATTTC[C/T]TTTGATCACATCCCTTACCTGCTCA
- Phenotype
-
MIM: 300871
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
FUNDC1
PubMed Links
Gene Details
- Gene
- FUNDC1
- Gene Name
- FUN14 domain containing 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_173794.3 |
|
Intron |
|
|
NP_776155.1 |
View Full Product Details