Product Details

SNP ID
rs11465553
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:52236960 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGCACATGGTGGATGACAGGGGTGA[C/T]GCAGGTGCAGCCAACAGTCACCAGC
Phenotype
MIM: 606496
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IL17F PubMed Links

Gene Details

Gene
IL17F
Gene Name
interleukin 17F
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_052872.3 600 Missense Mutation ATC,GTC I155V NP_443104.1
XM_011514276.1 600 Missense Mutation ATC,GTC I155V XP_011512578.1

View Full Product Details