Product Details

SNP ID
rs61734385
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:81813538 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGGCAAAGTGCTATGGGAGGAGC[A/G]GAACACCAGCAACCACAGGGCCTCA
Phenotype
MIM: 138033 MIM: 616514
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GCGR PubMed Links

Gene Details

Gene
GCGR
Gene Name
glucagon receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000160.4 1578 Missense Mutation CAG,CGG Q428R NP_000151.1
XM_006722277.1 1578 Missense Mutation CAG,CGG Q428R XP_006722340.1
XM_011523539.1 1578 Missense Mutation CAG,CGG Q395R XP_011521841.1
XM_017024446.1 1578 Missense Mutation CAG,CGG Q426R XP_016879935.1
XM_017024447.1 1578 Missense Mutation CAG,CGG Q306R XP_016879936.1
Gene
MCRIP1
Gene Name
MAPK regulated corepressor interacting protein 1
There are no transcripts associated with this gene.

View Full Product Details